Learn More
Mettler Toledo Generator electrode With Diaphragm
Catalog No. 01672603 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Coulometric Cell with Diaphragm
Catalog No. Type
01-672-603 Coulometric Cell with Diaphragm
1 options
Catalog No. 01-672-603 Supplier Mettler Toledo Supplier No. 31028002
Add to Cart
Add to Cart
Generate the titrant electrochemically in situ, with-diaphragm configuration releases directly into the titration solvent/sample mixture.
  • The digital plug-and-play generator electrode with a diaphragm maintains data integrity and enables easy tracking with unique serial numbers.
  • For use with METTLER TOLEDO EVA Coulometric Karl Fischer Titrators.
TRUSTED_SUSTAINABILITY
Specifications
Type Coulometric Cell with Diaphragm