Learn More
Mettler Toledo Generator electrode Without Diaphragm
Catalog No. 01672604 Shop All METTLER TOLEDO Products
Change view
Click to view available options
Type:
Coulometric Cell without Diaphragm
Catalog No. Type
01-672-604 Coulometric Cell without Diaphragm
1 options
Catalog No. 01-672-604 Supplier Mettler Toledo Supplier No. 31028003
Add to Cart
Add to Cart
Generate the titrant electrochemically in situ, without-diaphragm configuration releases directly into the titration solvent/sample mixture.
  • The digital plug-and-play generator electrode without a diaphragm maintains data integrity and enables easy tracking with unique serial numbers.
  • For use with METTLER TOLEDO EVA Coulometric Karl Fischer Titrators.
TRUSTED_SUSTAINABILITY
Specifications
Type Coulometric Cell without Diaphragm