Learn More
Pro Scientific Inc GENERATOR PROBE
SDP
Catalog No. NC9941346
Encompass_Preferred
PRO QUICK CONNECT GENERATORS,Generator Size:5 x 75mm,Bottom Type:Flat,Treatable Volume:0.03-5ml,They are manufactured from a high-quality 300 Series stainless steel and can be completely sterilized by hot air,autoclaving,flaming or chemical
Catalog No. NC9941346 Supplier Pro Scientific Inc Supplier No. 0205075
PRO QUICK CONNECT GENERATORS,Generator Size:5 x 75mm,Bottom Type:Flat,Treatable Volume:0.03-5ml,They are manufactured from a high-quality 300 Series stainless steel and can be completely sterilized by hot air,autoclaving,flaming or chemical