Learn More
Kam Controls Inc GENERATOR SOLUTION A 6/PK
Catalog No. NC9820145
Encompass
Kam Generator Solution A ,Component :Methanol -25%,Sulfur Dioxide :3%,Imidazole:-1%,Chloroform :25%,Hydriodic Acid :1%,Xylene:40%,Highly Flammable ,Wear Self-contained breathing apparatus and protecti
Catalog No. NC9820145 Supplier Kam Controls Inc Supplier No. 09004
Kam Generator Solution A ,Component :Methanol -25%,Sulfur Dioxide :3%,Imidazole:-1%,Chloroform :25%,Hydriodic Acid :1%,Xylene:40%,Highly Flammable ,Wear Self-contained breathing apparatus and protecti