Learn More
Kam Controls Inc GENERATOR SOLUTION C PK/6
Catalog No. NC9820146
Encompass
Kam Generator Solution C,Component :Diethanolaminhydrochlorid -50-60%,Methanol :35-45%,Diethanolaminsulfit :2-4%,Diethanolaminhydroiodide:1-3% ,Highly Flammable ,Toxic ,Extinguishing Media:Carbon Dioxide,Dry Chemical Powder or appropriate
Catalog No. NC9820146 Supplier Kam Controls Inc Supplier No. 09006
Kam Generator Solution C,Component :Diethanolaminhydrochlorid -50-60%,Methanol :35-45%,Diethanolaminsulfit :2-4%,Diethanolaminhydroiodide:1-3% ,Highly Flammable ,Toxic ,Extinguishing Media:Carbon Dioxide,Dry Chemical Powder or appropriate