Learn More
MSC Generic Duct Elbow, Galvanized, 5 in. ID, 45, 1 ea
Catalog No. 5000505473
Encompass
This galvanized duct elbow is designed for HVAC and ventilation systems, providing a 45-degree angle change in ductwork. It features a durable galvanized construction for longevity and resistance to corrosion.
  • Elbow fitting type is 45 degree
  • Inside diameter of 5 in.
  • Overall length of 7.86 in.
  • Galvanized material construction
  • Gage thickness of 22
Catalog No. 50-005-05473 Supplier MSC Supplier No. 03233194
This galvanized duct elbow is designed for HVAC and ventilation systems, providing a 45-degree angle change in ductwork. It features a durable galvanized construction for longevity and resistance to corrosion.
  • Elbow fitting type is 45 degree
  • Inside diameter of 5 in.
  • Overall length of 7.86 in.
  • Galvanized material construction
  • Gage thickness of 22