Learn More
Echelon Biosciences Research Labs GENERIC PHOSPHATASE ASSAY KIT
SDP
Catalog No. 50720426
Encompass
Echelons Malachite Green Assay Kit is designed for the in vitro measurement of lipid and protein phosphatase activity. The liberated phosphate forms a colored complex with molybdate/malachite green, which is quantified by reading the absorbance at 620 nm. The assay uses water-soluble lipid substrates so there is no need to make phospholipid vesicles. The assay is fast, convenient and offers a flexible non-radioactive alternative for measuring specific phosphatase activities. The Malachite Green Assay kit can measure as little as 50 pmoles of free phosphate and has a high level of accuracy in the range of 200 to 2,000 pmol of phosphate per assay point. The kit includes enough reagents for 192 samples, standards, and controls. This item is non returnable
This item is not returnable. View return policy
Catalog No. 50-720-426 Supplier Echelon Biosciences Research Labs Supplier No. K1500EAKIT
Add to Cart
Add to Cart
Echelons Malachite Green Assay Kit is designed for the in vitro measurement of lipid and protein phosphatase activity. The liberated phosphate forms a colored complex with molybdate/malachite green, which is quantified by reading the absorbance at 620 nm. The assay uses water-soluble lipid substrates so there is no need to make phospholipid vesicles. The assay is fast, convenient and offers a flexible non-radioactive alternative for measuring specific phosphatase activities. The Malachite Green Assay kit can measure as little as 50 pmoles of free phosphate and has a high level of accuracy in the range of 200 to 2,000 pmol of phosphate per assay point. The kit includes enough reagents for 192 samples, standards, and controls. This item is non returnable