Learn More
ORS Nasco GENESIS BLACK FR CLEAR UD
Catalog No. 502551578
Encompass_Preferred
Honeywell part# S3200. Honeywell Uvex Genesis Protective Safety Glasses Frame Color: Black Lens Type: Scratch-resistant Lens Tint: Clear Certifications/Compliance: ANSI Z87.1-2015 CSA Z94.3-2007 Military V0 Ballistic Test Description: Lifetime frame guarantee Recommended Applications: Safety
Catalog No. 50-255-1578 Supplier ORS Nasco Supplier No. 763S3200
Honeywell part# S3200. Honeywell Uvex Genesis Protective Safety Glasses Frame Color: Black Lens Type: Scratch-resistant Lens Tint: Clear Certifications/Compliance: ANSI Z87.1-2015 CSA Z94.3-2007 Military V0 Ballistic Test Description: Lifetime frame guarantee Recommended Applications: Safety