Learn More
MSC Uvex Genesis XC Safety Glasses, Polycarbonate, SCT-Low IR, Universal, 1 ea.
Catalog No. 5000509274
Encompass
These XC series safety glasses provide high-impact protection with an ergonomically balanced, multi-material frame. Designed for comfort and customization, they feature an extended 9-base wraparound lens that offers up to 20% more coverage around the eyes and cheeks. The adjustable, ratcheting temples allow for customized lens inclination, while soft, flexible nose fingers conform to various nasal profiles to minimize slippage.
  • Provides 99.9% UVA and UVB protection
  • Features dual-coating technology for anti-fog, anti-scratch, and anti-static performance
  • Includes soft, cushioned temple tips for all-day comfort
  • Utilizes soft elastomer browguard to diffuse and deflect impact
  • Compatible with prescription lens inserts ranging from +3.0 to -3.0 D
  • Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards
Catalog No. 50-005-09274 Supplier MSC Supplier No. 32311474
These XC series safety glasses provide high-impact protection with an ergonomically balanced, multi-material frame. Designed for comfort and customization, they feature an extended 9-base wraparound lens that offers up to 20% more coverage around the eyes and cheeks. The adjustable, ratcheting temples allow for customized lens inclination, while soft, flexible nose fingers conform to various nasal profiles to minimize slippage.
  • Provides 99.9% UVA and UVB protection
  • Features dual-coating technology for anti-fog, anti-scratch, and anti-static performance
  • Includes soft, cushioned temple tips for all-day comfort
  • Utilizes soft elastomer browguard to diffuse and deflect impact
  • Compatible with prescription lens inserts ranging from +3.0 to -3.0 D
  • Meets ANSI Z87.1-2015 and CSA Z94.3-2007 standards