Learn More
MSC Uvex Genesis XC Safety Glasses, Shade 5.0, HydroShield Anti-Fog, 1 ea
Catalog No. 5000508083
Encompass
These high-performance safety glasses feature a 9-base, wraparound green lens with a shade 5.0 IR filter designed for welding applications. Built with a semi-rimless black frame, they utilize multi-material technology to ensure a secure, customized fit for a variety of users. The integrated design provides extended cheek and side coverage, while the specialized lens coating enhances clarity and durability in demanding work environments.
  • Shade 5.0 IR filter lens for welding protection
  • HydroShield® anti-fog and anti-scratch lens coating
  • Soft elastomer brow guard deflects impact
  • Flexible nose fingers conform to different nasal profiles
  • Adjustable, ratcheting temples with cushioned tips for custom inclination
  • Extended wrap-around lens provides 20% more side coverage
  • Meets ANSI Z87.1+ and CSA Z94.3 safety standards
Catalog No. 50-005-08083 Supplier MSC Supplier No. 32311490
These high-performance safety glasses feature a 9-base, wraparound green lens with a shade 5.0 IR filter designed for welding applications. Built with a semi-rimless black frame, they utilize multi-material technology to ensure a secure, customized fit for a variety of users. The integrated design provides extended cheek and side coverage, while the specialized lens coating enhances clarity and durability in demanding work environments.
  • Shade 5.0 IR filter lens for welding protection
  • HydroShield® anti-fog and anti-scratch lens coating
  • Soft elastomer brow guard deflects impact
  • Flexible nose fingers conform to different nasal profiles
  • Adjustable, ratcheting temples with cushioned tips for custom inclination
  • Extended wrap-around lens provides 20% more side coverage
  • Meets ANSI Z87.1+ and CSA Z94.3 safety standards