Learn More
Thermo Scientific™ Fiber Optic Probes for GENESYS™
Catalog No. 14385391
Change view
Click to view available options
For Use With (Equipment):
GENESYS G5
GENESYS G5 and Evolution Pro
GENESYS G5, Evolution One Plus, and Evolution Pro spectrophtometers
Type:
Fiber Optic Coupler
Fiber Optic Dip Probe System
Fiber Optic Dip Probe, 10 mm
Tip for Dip Probe, 10 mm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. For Use With (Equipment) Type
14-385-391 GENESYS G5 and Evolution Pro Fiber Optic Dip Probe, 10 mm
14-385-385 GENESYS G5 Fiber Optic Dip Probe System
14-385-386 GENESYS G5 Fiber Optic Coupler
14-385-395 GENESYS G5, Evolution One Plus, and Evolution Pro spectrophtometers Tip for Dip Probe, 10 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 14-385-391 Supplier Thermo Scientific™ Supplier No. 840305000
Add to Cart
Add to Cart
Dip, tap and measure - eliminate liquid transfer and cuvettes from your workflow.
Make the most of every sample. Accessories for Thermo Scientific™ GENESYS™ UV-Vis spectrophotometers are designed to enable you to work smarter, not harder. We offer a wide variety of options to meet your measurement requirements. Cable-free, snap-in design allows for easy exchange of accessories.
Preserve sample integrity while minimizing sample carryover.
TRUSTED_SUSTAINABILITY
Specifications
Type Fiber Optic Dip Probe, 10 mm
For Use With (Equipment) GENESYS G5 and Evolution Pro