Description
Thermo Scientific™ GENESYS™ Smart QC Software can make your UV-Vis analysis smoother and easier. GENESYS Smart QC software provides a simple way to pass judgement on a sample being tested by UV-Vis spectroscopy, delivering a straightforward functionality not readily available from other manufacturers. With GENESYS Smart QC software, UV-Vis analysis methods can be customized to deliver simple pass/fail assessments based on your specific guidelines. This limits the risk of misinterpreting data and speeds the time to action by streamlining decision-making points for users at all levels . The size, simplicity, and cost-effective operation of a UV-Vis spectrophotometer and the range of tests available make it an ideal instrument for industrial QA/QC labs, especially those performing food and beverage analysis.
The GENESYS Smart QC software package takes a twofold approach: It utilizes PC-controlled method development, followed by locally controlled method deployment on the desired analytical instrument.
- An analyst can use a personal computer (PC) to custom design a set of equations and designate how the data obtained on the local spectrophotometer should be interpreted.
- Local control is intended to be as simple as possible, employing a straightforward, simple interface to limit the amount spectral interpretation required of the user.
The customized method can be transferred from a research director’s PC to a GENESYS UV-Vis Spectrophotometer in the lab, which allows the local operations to be performed by lab technicians regardless of familiarity with UV-Vis analytical methods. A typical result of the pre-programmed method could be as basic as a PASS/FAIL readout.
- Method development is ideal for setting quality control parameters, especially for ingredient control
- GENESYS Smart QC software can be used anywhere a simple instrument read-out with limited interpretation is required
Specifications
Specifications
| Description | GENESYS Smart QC software |
| Type | GENESYS Smart QC software |