Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

EDVOTEK™ Genetic Flightplan: Navigating Galapagos Finch Diversity with SYBR™ Safe
SDPSCI_ED

Catalog No. 01672278 Shop All EDVOTEK Products
Change view
Click to view available options
Class Size:
8 Lab Groups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Class Size
01-672-278 8 Lab Groups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-672-278 Supplier EDVOTEK™ Supplier No. 921S
Add to Cart
Add to Cart

In this Cholesterol Diagnostics with SYBR™ Safe experiment, a simulated genetic test for hypercholesterolemia is demonstrated.

  • Includes SYBR™ Safe DNA Stain, NOT FlashBlue™ DNA Stain
  • Introduces PCR-RFLP analysis to analyze specific nucleotide sequences
  • Understand how electrophoresis separates differently-sized molecules
  • Perform agarose gel electrophoresis
  • Meets NGSS HS DCI LS1.A, LS3.A and LS3.B, through the discussion of the structure and function of DNA and heredity and variance of traits
  • Group Size: For 8 Gels
  • Time Required: Complete in 45 minutes
  • Kit Includes: Instructions, Ready-to-Load QuickStrip™ DNA Samples, UltraSpec-Agarose™, Electrophoresis Buffer (50X), SYBR™ Safe DNA Stain, Practice Gel Loading Solution.
  • Storage: Room Temperature Stable. Storage of Ready-to-Load QuickStrip™ samples in the Refrigerator is Recommended.

Specifications

Grade 8 to Undergraduate
Class Size 8 Lab Groups
Product Type Genetics Concepts Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.