Learn More
Apexbio Technology LLC Geneticin, G-418 Sulfate, 108321-42-2, MFCD05664725, 1 g
SDP
Catalog No. 501150451
Encompass_Preferred
MF: C20H40N4O102H2SO4, MW: 692.71, Solubility: insoluble in DMSO; 240.4 mg/ml in H2O, Synonyms: Antibiotic G418. G-418 is an elongation inhibitor of 80 S ribosomes. G418 is an aminoglycoside antibiotic, it has a wide spectrum of activity and is active against bacteria, yeasts, fungi, algae, plant and even animal cells.
Catalog No. 50-115-0451 Supplier Apexbio Technology LLC Supplier No. A25131000
MF: C20H40N4O102H2SO4, MW: 692.71, Solubility: insoluble in DMSO; 240.4 mg/ml in H2O, Synonyms: Antibiotic G418. G-418 is an elongation inhibitor of 80 S ribosomes. G418 is an aminoglycoside antibiotic, it has a wide spectrum of activity and is active against bacteria, yeasts, fungi, algae, plant and even animal cells.