Learn More
Selleck Chemical LLC Geneticin (G418 Sulfate) 1g 108321-42-2
SDP
Catalog No. 501363278
Encompass_Preferred
Geneticin (G418 Sulfate), an aminoglycoside antibiotic, is an elongation inhibitor of 80 S ribosomes that blocks polypeptide synthesis by inhibiting the elongation step in both prokaryotic and eukaryotic cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-3278 Supplier Selleck Chemical LLC Supplier No. S30281G
Geneticin (G418 Sulfate), an aminoglycoside antibiotic, is an elongation inhibitor of 80 S ribosomes that blocks polypeptide synthesis by inhibiting the elongation step in both prokaryotic and eukaryotic cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.