Learn More
TOKU-E G418 Disulfate (Low Endotoxin), 108321-42-2, 1 g
Catalog No. 502551792
Encompass_Preferred
Geneticin Sulfate, aminoglycoside, selection antibiotic, endotoxin content ≤ 1EU/mg, C20H40N4O10 2H2SO4. Useful in cell culture, transfection.
Catalog No. 50-255-1792 Supplier TOKU-E Supplier No. G0485G
Geneticin Sulfate, aminoglycoside, selection antibiotic, endotoxin content ≤ 1EU/mg, C20H40N4O10 2H2SO4. Useful in cell culture, transfection.