Learn More
Medchemexpress LLC Geneticin sulfate; antibiotic G-418 sulfate | 108321-42-2 | 99.9% | 692.71 | 250 MG
SDP
Catalog No. 5000384397
Encompass_Preferred
G-418 disulfate, also known as Geneticin sulfate, is an aminoglycoside antibiotic. It functions by inhibiting protein synthesis in both eukaryotic and prokaryotic cells. This compound is widely utilized as a selective agent, particularly for eukaryotic cells in various biological applications.
  • Inhibits protein synthesis in eukaryotes and prokaryotes.
  • Commonly used as a selective agent for eukaryotic cells.
  • Interferes with protein synthesis.
  • Used for selection of mammalian cell lines possessing neomycin resistance.
  • Can be covalently bound to asialoorosomucoid (AsOR) for hepatocyte-specific targeting and toxicity.
  • Inhibitor of many pro- and eukaryotes.
Catalog No. 50-003-84397 Supplier Medchemexpress LLC Supplier No. 250MGHY17561R
G-418 disulfate, also known as Geneticin sulfate, is an aminoglycoside antibiotic. It functions by inhibiting protein synthesis in both eukaryotic and prokaryotic cells. This compound is widely utilized as a selective agent, particularly for eukaryotic cells in various biological applications.
  • Inhibits protein synthesis in eukaryotes and prokaryotes.
  • Commonly used as a selective agent for eukaryotic cells.
  • Interferes with protein synthesis.
  • Used for selection of mammalian cell lines possessing neomycin resistance.
  • Can be covalently bound to asialoorosomucoid (AsOR) for hepatocyte-specific targeting and toxicity.
  • Inhibitor of many pro- and eukaryotes.