Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Geneticin Sulfate, G-Biosciences
SCI_ED

Catalog No. S139823
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139823 25 g
S139776 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S139823 Supplier G-Biosciences Supplier No. RC856
Add to Cart
Add to Cart

Geneticin Sulfate (G-418) inhibits protein elongation by blocking polypeptide synthesis.

Eukaryotic cells that are stably transfected with neomycin resistance genes can be selected and maintained using this product. Similar in structure to Gentamicin, Neomycin, and Kanamycin.

Chemical Identifiers

Molecular Weight (g/mol) 692.71 g/mol

Specifications

Boiling Point 1012.1°C
Quantity 25 g
Percent Purity 98%
Grade Laboratory
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.