Learn More
3B Scientific™ Genetics Reproduction and Embryology Slide Set
SDPSCI_ED
Catalog No. S95014L Shop All 3B Scientific Products
Change view
Click to view available options
Quantity:
19
Catalog No. Quantity
S95014L 19
1 options
Catalog No. S95014L Supplier 3B Scientific™ Supplier No. W13404
Add to Cart
Add to Cart
19 microscope slides
  • 1(g). DNA and RNA stained in different colours, l.s. onion root tips
  • 2(e). Lilium, young anthers, meiosis, early prophase stage, t.s.
  • 3(e). Lilium, young anthers, diplotene stage, t.s.
  • 4(d). Lilium, ovary with embryosac t.s.
  • 5(d). Capsella bursa pastoris, l.s. of embryos
  • 6(h). Human chromosomes, spread in the metaphase stage, w.m.
  • 7(g). Lamp brush chromosomes
  • 8(e). Hydra with testis t.s.
  • 9(e). Hydra with ovaries t.s.
  • 10(f). Tapeworm (Taenia), mature proglottid, w.m.
  • 11(f). Ascaris, sec. of uteri showing maturation of ova
  • 12(e). Cockchafer (Melolontha), ovaries t.s.
  • 13(d). Frog (Rana), testis t.s. showing spermatogenesis
  • 14(f). Frog embryology: four cell stage t.s.
  • 15(f). Frog: morula stage l.s.
  • 16(f). Frog: neurula stage t.s.
  • 17(f). Chicken (Gallus) embryology: 24 hour t.s.
  • 18(f). Chicken embryology: 72 hour t.s.
  • 19(d). Mouse, uterus containing embryo t.s.
Specifications
Type Genetics
Quantity 19