Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Genexus™ Barcodes 1-32 HD

Catalog No. A40261
Encompass_Preferred
Change view
Click to view available options
Quantity:
32 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40261 32 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A40261 Supplier Ion Torrent™ Supplier No. A40261
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Content And Storage Store at room temperature.
Sample Type DNA, RNA
Sequencing Type Genome, DNA, and RNA Sequencing
For Use With (Equipment) Genexus™ Integrated Sequencer
For Use With (Application) Sequencing
Libraries Targeted Sequencing Library
No. of Reactions 32
Product Line Genexus
Product Type Barcode
Quantity 32 Reactions
Shipping Condition Room Temperature
Workflow Step Library Generation
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.