Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Genexus™ Total RNA Purification Kit

Catalog No. A45541
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
48
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A45541 48
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. A45541 Supplier Ion Torrent™ Supplier No. A45541
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A set of consumables that enables sequential, automated extraction, and quantitation of total from various specimen types including whole blood, fresh/frozen tissue (ex. Spleen, Pancreas), peripheral blood leukocytes (PBL/buffy coat), bone marrow, and cell lysates (ex. BMMC, PBMC, cell lines).

  • A set of consumables that enable sequential, automated extraction, and quantitation of total RNA from various specimen types
  • Specimen types include whole blood, fresh/frozen tissue (e.g., spleen, pancreas), peripheral blood leukocytes (PBL/buffy coat), bone marrow, and cell lysates (e.g., BMMC, PBMC, cell lines)
  • Kit supports four runs of up to 12 samples each, utilizing the proven MagMAX technology for extraction and Qubit technology for post-extraction nucleic acid quantitation
  • Consumables are prefilled with most reagents necessary to perform the extraction, so only a few pipetting steps are required per sample to set up the run
  • Simply remove the foil seal, add enzymes and samples, then place the consumables onto the instrument
  • Use this kit in conjunction with the Genexus Multisample DNA Purification Kit (A45540) for comprehensive nucleic acid extraction from the same sample(s)
  • Achieve consistent extraction results with MagMAX technology
  • Reduce hands-on setup time using pre-filled consumables
  • Maximize efficiency with automated Qubit quantitation
  • Experience rapid turnaround times of as little as two hours from lysate to quantified nucleic acid
  • For research use only, not for use in diagnostic procedures
TRUSTED_SUSTAINABILITY

Specifications

Format Pre-filled Reagent Plates
Sample Type Bone Marrow, Cell Lysates (ex. BMMC, PBMC, Cell Lines), Peripheral Blood Leukocytes, Tissue (Fresh or Frozen), Whole Blood
For Use With (Equipment) Genexus™ Purification System
For Use With (Application) Next Generation Sequencing
No. of Reactions 48
Output Purified, quantified Total RNA
Purification Method MagMax™ bead-based purification
Shipping Condition Ambient
Workflow Step Sample Purification and Quantification

For Research Use Only. Not for use in Diagnostic Procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.