Learn More
Ion Torrent™ Genexus™ Controls
Description
The Ion Torrent Genexus Controls kit provides library and template preparation controls for use with the Ion Torrent Genexus Integrated Sequencer. The kit offers greater experimental control and empowers you to assess sample quality, help save time and reagents, and maximize productivity. Each kit contains an Ion AmpliSeq-based Genexus Control Panel and a Genexus Control DNA, which can prepare up to 32 automated control libraries, as well as a Genexus Control Library, sufficient for four template preparations.
The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.
Related Links:
Specifications
Specifications
| Content And Storage | Sufficient for 32 DNA to sequencing reactions OR 4 library to sequencing reactions Store at -10 to -30°C. |
| Sample Type | DNA, RNA |
| Sequencing Type | Genome, DNA, and RNA Sequencing |
| For Use With (Equipment) | Genexus™ Integrated Sequencer |
| For Use With (Application) | Sequencing |
| Libraries | Targeted Sequencing Library |
| Product Line | Genexus |
| Product Type | Control |
| Quantity | 1 kit |
| Shipping Condition | Dry Ice |
| Show More |
Safety and Handling
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.