Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Genexus™ Dx Barcodes 1-32 HD

Catalog No. A54104
Encompass_Preferred
Change view
Click to view available options
No. of Reactions:
32 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Reactions
A54104 32 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A54104 Supplier Ion Torrent™ Supplier No. A54104
Add to Cart
Add to Cart

The Ion Torrent Genexus Dx Barcodes 1-32 HD kit provides one plate filled with 32 dried-down, single-use barcodes and covered with a pierceable foil seal.

The Ion Torrent Genexus Dx Barcodes 1-32 HD kit provides one plate filled with 32 dried-down, single-use barcodes and covered with a pierceable foil seal. The barcodes can be flexibly used in increments of 1 to 32, until all barcodes are exhausted.

These barcodes are used with the Genexus Dx Integrated Sequencer, which is an in vitro diagnostic (IVD) medical device intended to be used by trained personnel in a professional laboratory environment for automated, qualitative targeted sequencing of human DNA or RNA extracted from formalin-fixed, paraffin-embedded (FFPE) tumor tissue samples or plasma samples.

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at room temperature.
Sequencing Type Genome, DNA, and RNA Sequencing
For Use With (Equipment) Genexus™ Dx Integrated Sequencer
For Use With (Application) Sequencing
Libraries Targeted Sequencing Library
No. of Reactions 32 Reactions
Nucleic Acid Type DNA, RNA, and cfTNA* (*Not available in the U.S.)
Quantity 1 Ea.
What is included in Genexus Dx Barcodes 1-32 HD (Cat. No. A54104)?

Genexus Dx Barcodes 1-32 HD provides one plate filled with 32 lyophilized, single-use, dual barcodes (Barcodes HD 1-32) and covered with a pierceable foil seal, eliminating the risk of cross contamination between wells. Each plate supports up to 32 reactions.

Will the Genexus Dx Software notify me if the barcodes had been previously used?

When necessary, the software will alert the user during the run plan setup that not enough barcodes are available.

How long can the Genexus Dx Barcodes remain on the Genexus Dx platform?

The Genexus Dx Barcode plates can remain on the instrument deck until all barcodes are used. Once you remove the barcode plate, you cannot put it back on to use again as the instrument has no way of tracking barcode usage.

For In Vitro Diagnostic Use. Not available in all countries or regions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.