Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Genexus™ Dx GX5™ Chip and Genexus™ Coupler

Catalog No. A54106
Encompass_Preferred
Change view
Click to view available options
Quantity:
2 chips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A54106 2 chips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A54106 Supplier Ion Torrent™ Supplier No. A54106
Add to Cart
Add to Cart

The Ion Torrent™ Genexus™ Dx GX5 Chip and Genexus Coupler includes two GX5 Dx chips and two Genexus couplers, sufficient for two initializations of the Ion Torrent Genexus Dx Integrated Sequencer.

  • The Ion Torrent™ Genexus™ Dx GX5 Chip and Genexus Coupler includes two GX5 Dx chips and two Genexus couplers
  • Sufficient for two initializations of the Genexus Dx Integrated Sequencer
  • The Genexus Dx Integrated Sequencer is an in vitro diagnostic (IVD) medical device intended to be used by trained personnel in a professional laboratory environment
  • For automated, qualitative targeted sequencing of human DNA or RNA extracted from formalin-fixed, paraffin-embedded (FFPE) tumor tissue samples or plasma samples
  • Store at room temperature
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • 2 GX5 Dx chips
• 2 Genexus couplers

Store at room temperature.

For Use With (Equipment) Genexus™ Dx Integrated Sequencer
For Use With (Application) Sequencing
Product Line Genexus™ Dx
Product Type Chip and Coupler
Quantity 2 chips
Software 2 GX5 Dx chips
Shipping Condition Room Temperature
Source 2 Genexus couplers
How could I detect if my Genexus Dx GX5 Chip and Genexus Coupler pair is faulty?

If the number of sample reads is very low, and CF-1 metrics fail QC, there could be a potential issue with the chip/coupler.

How long can Genexus Dx GX5 Chip and Genexus Coupler remain on the Genexus Dx instrument?

The chip and coupler remain installed on the instrument until all lanes are used or up to 2 weeks, whichever comes first. Once a chip and coupler set is removed from the Genexus Dx platform, it can never be reused.

How many sequencing reads are possible with Genexus Dx GX5 Chip?

The Genexus Dx GX5 Chip generates 12-15 million sequencing reads per lane, or 48-60 million reads per chip. The four-lane chip design provides maximum flexibility for sample processing, enabling sequencing of one to four lanes per run, with the in-use stability to utilize any remaining lanes within two weeks of the initial run.

What is the function of the coupler in Genexus Dx GX5 Chip and Genexus Coupler?

The coupler allows the pipettor to dispense reagents into the desired chip lane and it shuttles the chip between the loading and sequencing area on the Genexus Dx platform. Every time you change the chip, you should change the coupler.

What is contained in Genexus Dx GX5 Chip and Genexus Coupler?

The Genexus Dx GX5 Chip and Genexus Coupler (Cat. No. A54106) contains two GX5 chips and two Genexus couplers, sufficient for up to eight sequencing runs with the Genexus Dx Integrated Sequencer.

For In Vitro Diagnostic Use. Not available in all countries or regions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.