Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ion Torrent™ Genexus™ Dx Pipette Tips
Description
- Kit contains 12 individually wrapped racks of 96 tips each, specially designed to fit into designated spaces on the deck of the Genexus Dx Integrated Sequencer
- The Ion Torrent Genexus Dx Integrated Sequencer is an in vitro diagnostic (IVD) medical device intended to be used by trained personnel in a professional laboratory environment
- Intended for automated, qualitative targeted sequencing of human DNA or RNA extracted from formalin-fixed, paraffin-embedded (FFPE) tumor tissue samples or plasma samples
Specifications
Specifications
| Content And Storage | Store at room temperature. |
| Sequencing Type | Genome, DNA, and RNA Sequencing |
| For Use With (Equipment) | Genexus™ Dx Integrated Sequencer |
| For Use With (Application) | Sequencing |
| Product Line | Genexus™ Dx |
| Product Type | Pipette Tips |
| Quantity | 12 x 96 Tips |
| Shipping Condition | Room Temperature |
Frequently Asked Questions (FAQs)
How many pipette tips do I get with Genexus Dx Pipette Tips (Cat. No. A54105)?
12 racks of pipette tips are provided, containing 96 tips per rack. The pipette tips can be stored at room temperature.
For In Vitro Diagnostic Use. Not available in all countries or regions.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?