Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Genexus™ Library Strips 1 and 2-HD

Catalog No. A40255
Encompass_Preferred
Change view
Click to view available options
Quantity:
32 Reactions
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40255 32 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40255 Supplier Ion Torrent™ Supplier No. A40255
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

The Ion Torrent Genexus Library Strips 1 and 2-HD kit for Ion AmpliSeq HD-based panels provides eight each of the strips required to perform automated library preparation using the Ion Torrent Genexus Integrated Sequencer.

The Ion Torrent Genexus Library Strips 1 and 2-HD kit for Ion AmpliSeq HD-based panels provides eight each of the strips required to perform automated library preparation using the Ion Torrent Genexus Integrated Sequencer. This kit, combined with Ion Torrent Genexus Barcodes 1-32 HD, supports flexible, fully automated preparation and normalization of up to 32 single-pool Ion AmpliSeq HD libraries.

As part of the Genexus Integrated Sequencer workflow, the Genexus Library Strips 1 and 2-HD kit:

  • Automates library preparation for 200–400 base-read Ion AmpliSeq HD libraries
  • Supports intake variability of clinical research samples in a cost-effective manner
  • Maximizes flexibility with the ability prepare 1 to 32 single-pool libraries per run
  • Enables parallel processing of up to four compatible assays in a single run
  • Delivers fast and simple instrument setup, with less than five minutes of hands-on time

The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.

Related Links:

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage • Genexus Strip 2-HD, store at -10 to -30°C
• Genexus Strip 1, store at 2–8°C
Sample Type DNA, RNA
Sequencing Type Genome, DNA, and RNA Sequencing
For Use With (Equipment) Genexus™ Integrated Sequencer
For Use With (Application) Sequencing
Libraries Targeted Sequencing Library
No. of Reactions 32
Product Line Genexus
Product Type Library Strips 1 and 2-HD
Quantity 32 Reactions
Shipping Condition Approved for shipment on Wet or Dry Ice
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.