Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ion Torrent™ Genexus™ Pipette Tips

Catalog No. A40266
Encompass_Preferred
Change view
Click to view available options
Quantity:
12 x 96 Tips
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A40266 12 x 96 Tips
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A40266 Supplier Ion Torrent™ Supplier No. A40266
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

The Ion Torrent Genexus Pipette Tips kit contains 12 individually wrapped racks of 96 tips each, specially designed to fit into designated spaces on the deck of the Genexus Integrated Sequencer.

The Ion Torrent Genexus Pipette Tips kit contains 12 individually wrapped racks of 96 tips each, specially designed to fit into designated spaces on the deck of the Genexus Integrated Sequencer. The lid and wrapping keep the tips free of dust and contaminants prior to use, and the yellow base protects the tip from contacting surfaces during handling. Each run utilizes 2–6 racks of pipette tips, depending on configuration.

The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.

Related Links:

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 12 racks of 96 tips
Sample Type DNA, RNA
Sequencing Type Genome, DNA, and RNA Sequencing
For Use With (Equipment) Genexus™ Integrated Sequencer
For Use With (Application) Sequencing
Libraries Targeted Sequencing Library
Product Line Genexus
Product Type Pipette Tip
Quantity 12 x 96 Tips
Shipping Condition Room Temperature
Workflow Step Library Generation
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.