Learn More
Ion Torrent™ Genexus™ Primer Pool Tubes
Description
The Ion Torrent Genexus Primer Pool Tubes kit contains 50 empty assemblies of the custom tubes required to deliver primer oligos into the automated library preparation reaction. Each assembly is serialized for traceability and can be used with Ion AmpliSeq and Ion AmpliSeq HD-based assays. Pipette an aliquot of panel primers from a custom or AmpliSeq-on-Demand panel in order to run the assay on the Genexus Integrated Sequencer.
The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.
Related Links:
Specifications
Specifications
| Sample Type | DNA, RNA |
| Sequencing Type | Genome, DNA, and RNA Sequencing |
| For Use With (Equipment) | Genexus™ Integrated Sequencer |
| For Use With (Application) | Sequencing |
| Libraries | Targeted Sequencing Library |
| No. of Reactions | 200 |
| Product Line | Genexus |
| Product Type | Primer Pool Tube |
| Quantity | 200 Reactions |
| Shipping Condition | Room Temperature |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.