Learn More
Ion Torrent™ Genexus™ Sequencing Kit

Description
The Ion Torrent Genexus Sequencing Kit provides the materials required to perform two initializations and automated sequencing of two multilane chips using the Ion Torrent Genexus Integrated Sequencer. These innovative sequencing consumables are stable on the instrument for up to two weeks after initialization, allowing flexibility to use the chip lanes according to sample throughput needs. Genexus Bottles 1, 2, and 3 are shelf-stable and the Genexus Cartridge has a minimized footprint, limiting the required storage space at -20°C.
As part of the Genexus Integrated Sequencer workflow, the Genexus Sequencing Kit:
- Automates sequencing for 200–400 base-read Ion AmpliSeq or Ion AmpliSeq HD libraries
- Supports intake variability of clinical research samples in a cost-effective manner
- Maximizes flexibility with the ability to sequence one to four chip lanes per run
- Enables parallel processing of up to four compatible assays in a single run
- Delivers fast and simple instrument setup, with less than five minutes of hands-on time
The Ion Torrent Genexus Integrated Sequencer automates library preparation, sequencing, analysis, and reporting in a hands-off, walkaway workflow to deliver results in a single day. The innovative, multilane sequencing chip is designed to enable cost-effective processing of 1 to 32 single pool libraries.
Related Links:
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
| Content And Storage | • Genexus Cartridge, store at -10 to -30°C • Genexus Bottle 2, store at room temperature • Genexus Bottles 1 and 3, store at room temperature |
| Format | Kit |
| For Use With (Equipment) | Genexus™ Integrated Sequencer |
| For Use With (Application) | Comparative Sequencing (Somatic Mutations - 10:50) |
| Product Line | Genexus |
| Product Type | Sequencing Kit |
| Quantity | 2 Runs |
| Shipping Condition | Qualified for Ambient or Wet Ice |
For Research Use Only. Not for use in diagnostic procedures.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.