Learn More
Beckman Coulter GENFIND V3 REAGENT KIT - 384 P
Catalog No. NC2597946
Encompass
  • High-throughput genomic DNA isolation kit
  • Utilizes SPRI paramagnetic bead-based technology
  • Efficient recovery of high-quality DNA
  • Compatible with various sample types including whole blood
  • Supports multiple anticoagulants including citrate and EDTA
  • 384 preps for 200 µL samples, 192 preps for 400 µL samples
  • High quality, intact genomic DNA output
  • Applicable for PCR, genotyping, and Next-Generation Sequencing
  • Manual and automated workflow options available
  • Compatible with Beckman Coulter Biomek liquid handlers
  • Liquid format for ease of use
  • High throughput capability
  • Research use only
  • Minimizes hands-on time and pipetting errors
  • High DNA recovery rate with reduced PCR inhibitors
Catalog No. NC2597946 Supplier Beckman Coulter Supplier No. C34881
  • High-throughput genomic DNA isolation kit
  • Utilizes SPRI paramagnetic bead-based technology
  • Efficient recovery of high-quality DNA
  • Compatible with various sample types including whole blood
  • Supports multiple anticoagulants including citrate and EDTA
  • 384 preps for 200 µL samples, 192 preps for 400 µL samples
  • High quality, intact genomic DNA output
  • Applicable for PCR, genotyping, and Next-Generation Sequencing
  • Manual and automated workflow options available
  • Compatible with Beckman Coulter Biomek liquid handlers
  • Liquid format for ease of use
  • High throughput capability
  • Research use only
  • Minimizes hands-on time and pipetting errors
  • High DNA recovery rate with reduced PCR inhibitors