Learn More
Research Products International Corp Royal Genie Blotter with Platinized Titanium Anode, 20 x 20 cm
SDP
Catalog No. 50212273
Encompass
Research laboratories worldwide use Genie Blotters for rapid and accurate electrophoretic transfers. The unique design closely positions the plate electrodes to produce high field strengths(in volts/cm) with low voltage power sources. Superb Western Blots can be obtained in as little as 30 minutes at 24 volts. Northern and Southern blots in as little as 60-90 minutes.
GENIE also features an easy to load cassette tray for submerged bubble-free assembly. Slide the loaded tray into the tray holder and stand unit upright. The vertical operation eliminates bubble accumulation, gel slumping and burned blots. Supplied complete with cassette tray, tray holder, electrodes, Scotch-Brite pads, bubble screens and complete instructions.
Catalog No. 50-212-273 Supplier Research Products International Corp Supplier No. 144021
Research laboratories worldwide use Genie Blotters for rapid and accurate electrophoretic transfers. The unique design closely positions the plate electrodes to produce high field strengths(in volts/cm) with low voltage power sources. Superb Western Blots can be obtained in as little as 30 minutes at 24 volts. Northern and Southern blots in as little as 60-90 minutes.
GENIE also features an easy to load cassette tray for submerged bubble-free assembly. Slide the loaded tray into the tray holder and stand unit upright. The vertical operation eliminates bubble accumulation, gel slumping and burned blots. Supplied complete with cassette tray, tray holder, electrodes, Scotch-Brite pads, bubble screens and complete instructions.