Learn More
Research Products International Corp Genie Cell Disrupter, Features Patented Multi-Direction Action
SDP
Catalog No. 501977023
Encompass
Ideal for applications requiring cell disruptions and homogenization of yeast cells. Genie Disruptor features patented multi-direction action that simultaneously agitates and vortexes samples at high speeds. Produces greater yields in less time when used with glass beads. Features 0-15 minute auto-off timer or continuous operation. Suitable for use in cold rooms or incubators. Heavy duty metal casting and stainless steel construction prevents unit from moving during operation. Supplied with tube holder for 12 x 1.5ml micro-tubes. Two year warranty.
Catalog No. 50-197-7023 Supplier Research Products International Corp Supplier No. 195575
Ideal for applications requiring cell disruptions and homogenization of yeast cells. Genie Disruptor features patented multi-direction action that simultaneously agitates and vortexes samples at high speeds. Produces greater yields in less time when used with glass beads. Features 0-15 minute auto-off timer or continuous operation. Suitable for use in cold rooms or incubators. Heavy duty metal casting and stainless steel construction prevents unit from moving during operation. Supplied with tube holder for 12 x 1.5ml micro-tubes. Two year warranty.