Learn More
OHAUS Genie SS Beads
Catalog No. 01804413 Shop All OHAUS Products
Change view
Click to view available options
Diameter (Metric):
2.4 mm
3.2 mm
5 mm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric)
01-804-413 5 mm
01-804-344 2.4 mm
01-804-402 3.2 mm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-804-413 Supplier OHAUS Supplier No. 83041298
Add to Cart
Edge
Add to Cart
Bead balls are made of Type 316 stainless steel.
  • For bead beating applications.
  • Use with Bead Genie and Disruptor Genie™.
  • Can be cleaned and reused.
TRUSTED_SUSTAINABILITY
Specifications
Product Type Beads
Diameter (Metric) 5 mm
Model Bead Genie
Packaging Quantity 100 Pk.