Learn More
Cayman Chemical GenInthIocIn A 2.5mg
SDP
Catalog No. 502941705
Encompass_Preferred
A cyclic thiopeptide bacterial metabolite; active against a variety of Gram-positive bacteria (MICs = 0.2-4 UG/ml), the Gram-negative bacterium C. violaceum (MIC = 19 UG/ml), and the fungus M. hiemalis (MIC = 38 UG/ml); induces tipA promoter transcription with a minimum induction concentration of 1.2 ng/ml
Catalog No. 50-294-1705 Supplier Cayman Chemical Supplier No. 281252.5 MG
Add to Cart
Add to Cart
A cyclic thiopeptide bacterial metabolite; active against a variety of Gram-positive bacteria (MICs = 0.2-4 UG/ml), the Gram-negative bacterium C. violaceum (MIC = 19 UG/ml), and the fungus M. hiemalis (MIC = 38 UG/ml); induces tipA promoter transcription with a minimum induction concentration of 1.2 ng/ml