Learn More
Medchemexpress LLC Genipin 1-β-D-gentiobioside | 29307-60-6 | 10 MM 1 ML
SDP
Catalog No. 5000420563
Encompass_Preferred
Genipin 1-β-D-gentiobioside is an abundant and bioactive iridoid glycoside found in Gardenia jasminoides Ellis. It exhibits hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic activities. This product is for research use only.
  • Abundant and bioactive iridoid glycoside
  • Exhibits hepatoprotective activity
  • Exhibits anti-inflammatory activity
  • Exhibits antioxidant activity
  • Exhibits antithrombotic activity
  • Suitable for research use
Catalog No. 50-004-20563 Supplier Medchemexpress LLC Supplier No. HYN209410MM/1ML
Genipin 1-β-D-gentiobioside is an abundant and bioactive iridoid glycoside found in Gardenia jasminoides Ellis. It exhibits hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic activities. This product is for research use only.
  • Abundant and bioactive iridoid glycoside
  • Exhibits hepatoprotective activity
  • Exhibits anti-inflammatory activity
  • Exhibits antioxidant activity
  • Exhibits antithrombotic activity
  • Suitable for research use