Learn More
Medchemexpress LLC Genipin 1-β-D-gentiobioside | 29307-60-6 | 25 MG
SDP
Catalog No. 5000422077
Encompass_Preferred
Genipin 1-β-D-gentiobioside is one of the most abundant and bioactive iridoid glycosides found in *Gardenia jasminoides Ellis*. It is intended for research use only and is not sold to patients.
  • Exhibits hepatoprotective activity.
  • Exhibits anti-inflammatory activity.
  • Exhibits antioxidant activity.
  • Exhibits antithrombotic activity.
  • Has a purity of 99.73%.
  • Appears as a white to light yellow solid.
Catalog No. 50-004-22077 Supplier Medchemexpress LLC Supplier No. 25MGHYN2094
Genipin 1-β-D-gentiobioside is one of the most abundant and bioactive iridoid glycosides found in *Gardenia jasminoides Ellis*. It is intended for research use only and is not sold to patients.
  • Exhibits hepatoprotective activity.
  • Exhibits anti-inflammatory activity.
  • Exhibits antioxidant activity.
  • Exhibits antithrombotic activity.
  • Has a purity of 99.73%.
  • Appears as a white to light yellow solid.