Learn More
Medchemexpress LLC Genipin 1-β-D-gentiobioside | 29307-60-6 | 5 MG
SDP
Catalog No. 5000422042
Encompass_Preferred
Genipin 1-β-D-gentiobioside is one of the most abundant and bioactive iridoid glycosides in Gardenia jasminoides Ellis. It possesses hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic activities.
  • Derived from Gardenia jasminoides Ellis
  • Possesses hepatoprotective activity
  • Exhibits anti-inflammatory activity
  • Shows antioxidant activity
  • Has antithrombotic activity
  • For research use only
Catalog No. 50-004-22042 Supplier Medchemexpress LLC Supplier No. HYN20945MG
Genipin 1-β-D-gentiobioside is one of the most abundant and bioactive iridoid glycosides in Gardenia jasminoides Ellis. It possesses hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic activities.
  • Derived from Gardenia jasminoides Ellis
  • Possesses hepatoprotective activity
  • Exhibits anti-inflammatory activity
  • Shows antioxidant activity
  • Has antithrombotic activity
  • For research use only