Learn More
Cayman Chemical GenIpIn 1-gentIobIosIde 1mg
SDP
Catalog No. 502913344
Encompass_Preferred
An iridoid glycoside; inhibits AChE by 60.23% at 25 UG/ml; is cytotoxic to BRL 3A rat liver fibroblasts from 50-500 UG/ml; inhibits the germination of G. jasminoides and N. offinicale seeds and the growth of B. rapa roots; prevents scopolamine-induced increases in escape latency in the Morris water maze in mice at 2.5 mg/kg
Catalog No. 50-291-3344 Supplier Cayman Chemical Supplier No. 356211 MG
Add to Cart
Add to Cart
An iridoid glycoside; inhibits AChE by 60.23% at 25 UG/ml; is cytotoxic to BRL 3A rat liver fibroblasts from 50-500 UG/ml; inhibits the germination of G. jasminoides and N. offinicale seeds and the growth of B. rapa roots; prevents scopolamine-induced increases in escape latency in the Morris water maze in mice at 2.5 mg/kg