Learn More
Medchemexpress LLC Genipin 1-gentiobioside | 29307-60-6 | 99.7% | 550.51 g/mol | C23H34O15 | 10 MG
SDP
Catalog No. 5000296662
Encompass_Preferred
Genipin 1-gentiobioside is an iridoid glycoside isolated from Gardenia jasminoides, provided as a research reagent for biological and pharmacological studies. It is commonly used in bioactivity screening and analytical applications due to its reported hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic properties.
  • High purity suitable for analytical and bioassay applications.
  • Reported bioactivities including hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic effects.
  • Characterized by CAS 29307-60-6, molecular formula C23H34O15, molecular weight 550.51 g/mol.
  • Supplied in small vials to support screening and method development workflows.
Catalog No. 50-002-96662 Supplier Medchemexpress LLC Supplier No. HYN209410MG
Genipin 1-gentiobioside is an iridoid glycoside isolated from Gardenia jasminoides, provided as a research reagent for biological and pharmacological studies. It is commonly used in bioactivity screening and analytical applications due to its reported hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic properties.
  • High purity suitable for analytical and bioassay applications.
  • Reported bioactivities including hepatoprotective, anti-inflammatory, antioxidant, and antithrombotic effects.
  • Characterized by CAS 29307-60-6, molecular formula C23H34O15, molecular weight 550.51 g/mol.
  • Supplied in small vials to support screening and method development workflows.