Learn More
Enzo Life Sciences Genipin (100mg). CAS: 6902-77-8
SDP
Catalog No. 502008874
Encompass_Preferred
Cell permeable inhibitor of uncoupling protein 2 (UCP2). Increases glucose-stimulated insulin secretion, mitochondrial membrane potential and ATP levels in pancreatic island cells. Induces apoptosis. Protein cross-linking agent. Anti-inflammatory and anti-angiogenic. Alternative name: (+)-Genipin, Methyl (1S,2R,6S)-2-hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate. Purity: ≥98% (HPLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in DMSO (50mg/ml), dimethyl formamide (25mg/ml), 100% ethanol (5mg/ml), methanol or ethyl acetate; insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8874 Supplier Enzo Life Sciences Supplier No. ALX350383M100
Cell permeable inhibitor of uncoupling protein 2 (UCP2). Increases glucose-stimulated insulin secretion, mitochondrial membrane potential and ATP levels in pancreatic island cells. Induces apoptosis. Protein cross-linking agent. Anti-inflammatory and anti-angiogenic. Alternative name: (+)-Genipin, Methyl (1S,2R,6S)-2-hydroxy-9-(hydroxymethyl)-3-oxabicyclo[4.3.0]nona-4,8-diene-5-carboxylate. Purity: ≥98% (HPLC). Appearance: White to off-white crystalline solid. Solubility: Soluble in DMSO (50mg/ml), dimethyl formamide (25mg/ml), 100% ethanol (5mg/ml), methanol or ethyl acetate; insoluble in water. Long Term Storage: -20°C.