Learn More
TARGETMOL CHEMICALS INC Genipin 100MG
SDP
Catalog No. 502877472
Encompass_Preferred
Also available in 1 mL, 20 mg, 50 mg and bulk. Please contact Fisher for quotes. Genipin ((+)-Genipin), an active aglycone derived from an iridoid glycoside called geniposide, is found in the fruit of Gardenia jasminoides Ellis. Purity 99.74%
Catalog No. 50-287-7472 Supplier TARGETMOL CHEMICALS INC Supplier No. T2210100MG
Also available in 1 mL, 20 mg, 50 mg and bulk. Please contact Fisher for quotes. Genipin ((+)-Genipin), an active aglycone derived from an iridoid glycoside called geniposide, is found in the fruit of Gardenia jasminoides Ellis. Purity 99.74%