Learn More
AdipoGen Genipin, 6902-77-8, MFCD00888600, 100 mg
SDP
Catalog No. 501146398
Encompass_Preferred
MF: C11H14O5, MW: 226.2, ≥98%, Appearance: White to off-white solid. Cell permeable inhibitor of uncoupling protein 2. Increases glucose-stimulated insulin secretion, mitochondrial membrane potential and ATP levels in pancreatic island cells. Anticancer, antimetastatic and anti-angiogenic agent. Increases Bax/Bcl-2 ratio and induces PARP cleavage and caspase-3- and caspase-9-mediated apoptosis in non-small cell lung cancer (NSCLC) cells. Sensitizes multidrug-resistant cancer cells to cytotoxic agents through UCP2 inhibition. Suppresses the tumor promoting function of UCP2 in a cellular model of Warburg effect.
Catalog No. 50-114-6398 Supplier AdipoGen Supplier No. AGCN20481M100
MF: C11H14O5, MW: 226.2, ≥98%, Appearance: White to off-white solid. Cell permeable inhibitor of uncoupling protein 2. Increases glucose-stimulated insulin secretion, mitochondrial membrane potential and ATP levels in pancreatic island cells. Anticancer, antimetastatic and anti-angiogenic agent. Increases Bax/Bcl-2 ratio and induces PARP cleavage and caspase-3- and caspase-9-mediated apoptosis in non-small cell lung cancer (NSCLC) cells. Sensitizes multidrug-resistant cancer cells to cytotoxic agents through UCP2 inhibition. Suppresses the tumor promoting function of UCP2 in a cellular model of Warburg effect.