Learn More
Medchemexpress LLC HY-17389 100mg , Genipin (+)-Genipin CAS:6902-77-8 Purity:98%
SDP
Catalog No. 501493520
Encompass_Preferred
Medchemexpress, HY-17389 100mg Genipin (+)-Genipin CAS:6902-77-8 Formula:C11H14O5 Purity:98% Genipin is a cell permeable inhibitor of uncoupling protein 2 (UCP2). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3520 Supplier Medchemexpress LLC Supplier No. HY17389100MG
 Restricted Item
Add to Cart
Add to Cart
Medchemexpress, HY-17389 100mg Genipin (+)-Genipin CAS:6902-77-8 Formula:C11H14O5 Purity:98% Genipin is a cell permeable inhibitor of uncoupling protein 2 (UCP2). Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.