Learn More
Apexbio Technology LLC Genipin 6902-77-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063871
Encompass_Preferred
Genipin (CAS 6902-77-8) is a natural small molecule isolated from Gardenia fruit traditionally employed in Chinese medicine for managing type 2 diabetes symptoms It demonstrates antiviral activity against hepatitis C virus exhibiting an IC50 of 88 41 M In vitro studies show Genipin enhances glucose uptake in C2C12 myotubes by increasing GLUT4 translocation intracellular ATP and calcium levels via ATP-sensitive potassium channel closure with dependence on PI3-K and calcium signaling pathways Additionally in rat liver models Genipin enhances Mrp2-mediated bile secretion and transporter localization without altering Mrp2 mRNA
Catalog No. 50-000-63871 Supplier Apexbio Technology LLC Supplier No. N24085.1
Genipin (CAS 6902-77-8) is a natural small molecule isolated from Gardenia fruit traditionally employed in Chinese medicine for managing type 2 diabetes symptoms It demonstrates antiviral activity against hepatitis C virus exhibiting an IC50 of 88 41 M In vitro studies show Genipin enhances glucose uptake in C2C12 myotubes by increasing GLUT4 translocation intracellular ATP and calcium levels via ATP-sensitive potassium channel closure with dependence on PI3-K and calcium signaling pathways Additionally in rat liver models Genipin enhances Mrp2-mediated bile secretion and transporter localization without altering Mrp2 mRNA