Learn More
Selleck Chemical LLC Genipin S2412-1g
SDP
Catalog No. 503111757
Encompass_Preferred
Genipin(( )-Genipin) is an active aglycone derived from an iridoid glycoside called geniposide which is found in the fruit of Gardenia jasminoides Ellis
Catalog No. 50-311-1757 Supplier Selleck Chemical LLC Supplier No. S24121G
Genipin(( )-Genipin) is an active aglycone derived from an iridoid glycoside called geniposide which is found in the fruit of Gardenia jasminoides Ellis