Learn More
Medchemexpress LLC Genipin | 6902-77-8 | 99.8% | 226.23 | 1 ML
SDP
Catalog No. 5000383949
Encompass_Preferred
Genipin is a natural crosslinking reagent derived from Gardenia jasminoides Ellis fruits. It inhibits UCP2 (uncoupling protein 2) in cells and is suitable for various research applications, including type 2 diabetes research.
  • Inhibits UCP2 (uncoupling protein 2) in cells
  • Modulates proteins
  • Antitumor activity
  • Anti-inflammation activity
  • Immunosuppression
  • Antithrombosis
  • Protects hippocampal neurons
Catalog No. 50-003-83949 Supplier Medchemexpress LLC Supplier No. HY1738910MM/1ML
Genipin is a natural crosslinking reagent derived from Gardenia jasminoides Ellis fruits. It inhibits UCP2 (uncoupling protein 2) in cells and is suitable for various research applications, including type 2 diabetes research.
  • Inhibits UCP2 (uncoupling protein 2) in cells
  • Modulates proteins
  • Antitumor activity
  • Anti-inflammation activity
  • Immunosuppression
  • Antithrombosis
  • Protects hippocampal neurons