Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Genipin | 6902-77-8 | MFCD00888600 | >98.0% | 226.23 | C11H14O5 | 10 MG
SDP

Catalog No. 5000299178
Encompass_Preferred

Genipin is a natural crosslinking reagent derived from Gardenia jasminoides fruits, commonly used in biochemical and materials research. It inhibits uncoupling protein 2 (UCP2) and displays bioactivities including protein modulation, anti-inflammatory, antitumor, immunosuppressive, and neuroprotective effects. Supplied as a high-purity solid suitable for analytical and research applications.

  • Natural crosslinking reagent derived from Gardenia jasminoides.
  • Inhibits uncoupling protein 2 (UCP2).
  • Useful in research areas such as autophagy, metabolic, and neurological disease.
  • Purity >98.0%.
  • Molecular weight 226.23 g/mol.
  • Light yellow to yellow solid powder appearance.
  • Storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in small pack sizes including 10 mg for analytical use.

Catalog No. 50-002-99178 Supplier Medchemexpress LLC Supplier No. HY17389R10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Genipin is a natural crosslinking reagent derived from Gardenia jasminoides fruits, commonly used in biochemical and materials research. It inhibits uncoupling protein 2 (UCP2) and displays bioactivities including protein modulation, anti-inflammatory, antitumor, immunosuppressive, and neuroprotective effects. Supplied as a high-purity solid suitable for analytical and research applications.

  • Natural crosslinking reagent derived from Gardenia jasminoides.
  • Inhibits uncoupling protein 2 (UCP2).
  • Useful in research areas such as autophagy, metabolic, and neurological disease.
  • Purity >98.0%.
  • Molecular weight 226.23 g/mol.
  • Light yellow to yellow solid powder appearance.
  • Storage: powder -20°C (3 years) or 4°C (2 years).
  • Available in small pack sizes including 10 mg for analytical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.