Learn More
Medchemexpress LLC Genipin | 6902-77-8 | MFCD00888600 | 99.8% | 226.23 | 25 MG
SDP
Catalog No. 5000383978
Encompass_Preferred
Genipin ((+)-Genipin) is a natural crosslinking reagent derived from Gardenia jasminoides Ellis fruits. It inhibits UCP2 (uncoupling protein 2) in cells. This compound exhibits various bioactivities, making it suitable for a range of research applications.
  • Modulates proteins
  • Antitumor properties
  • Anti-inflammation capabilities
  • Immunosuppressive effects
  • Antithrombosis activity
  • Protects hippocampal neurons
  • Used for type 2 diabetes research
Catalog No. 50-003-83978 Supplier Medchemexpress LLC Supplier No. 25MGHY17389
Genipin ((+)-Genipin) is a natural crosslinking reagent derived from Gardenia jasminoides Ellis fruits. It inhibits UCP2 (uncoupling protein 2) in cells. This compound exhibits various bioactivities, making it suitable for a range of research applications.
  • Modulates proteins
  • Antitumor properties
  • Anti-inflammation capabilities
  • Immunosuppressive effects
  • Antithrombosis activity
  • Protects hippocampal neurons
  • Used for type 2 diabetes research