Learn More
Cayman Chemical Genipin, 6902-77-8, 50 mg
SDP
Supplier:  Cayman Chemical 1001062250
Encompass_Preferred
A protein cross-linking agent isolated from G. jasminoides Ellis; inhibits UCP2 activity; 5 µM genipin increases glucose-stimulated insulin secretion in isolated pancreatic islets chronically exposed to high levels of glucose; induces apoptosis in FaO rat hepatoma cells and human hepatocarcinoma Hep3B cells
Catalog No. NC0063949