Learn More
Abcam Genipin, Cell-permeable uncoupling protein 2 inhibitor, 100MG
Catalog No. 502706949
Encompass_Preferred
MW 226.23 g/mol, Purity >99%. Cell-permeable uncoupling protein 2 (UCP2) inhibitor. Anti-inflammatory, anti-angiogenic and pro-apototic effects. Acts as a protein cross-linker.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6949 Supplier Abcam Supplier No. AB141049100MG
MW 226.23 g/mol, Purity >99%. Cell-permeable uncoupling protein 2 (UCP2) inhibitor. Anti-inflammatory, anti-angiogenic and pro-apototic effects. Acts as a protein cross-linker.
The product is subject to the following: Abcam Restricted Use Statement